Learn More
Emma Safety Footwear BAS Safety Shoes
Product Code. 15938841
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15938841 37 1 pair
15948841 38 1 pair
15958841 39 1 pair
15968841 40 1 pair
15978841 41 1 pair
15988841 42 1 pair
15998841 43 1 pair
15908851 44 1 pair
15918851 45 1 pair
15928851 46 1 pair
15938851 47 1 pair
15948851 48 1 pair
15958851 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15938841 Supplier Emma Safety Footwear Supplier No. MM70084837
Add to basket
Add to basket
All purpose safety boot. The outsole repels acid and oil like an umbrella repels water. Strong ankle supporting upper fits the foot snugly and comfortably.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011
Closure Type Laces
Color Black
Design BAS
Liner Material Hydro-Tec™ Sanitized Silver
Material Leather
Size 37
Toe Material Steel