Learn More
Emma Safety Footwear BAS XD Safety Shoes
Product Code. 15998871
Change view
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15998871 49 1 pair
15978861 37 1 pair
15988861 38 1 pair
15998861 39 1 pair
15908871 40 1 pair
15918871 41 1 pair
15928871 42 1 pair
15938871 43 1 pair
15948871 44 1 pair
15958871 45 1 pair
15968871 46 1 pair
15978871 47 1 pair
15988871 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 15998871 Supplier Emma Safety Footwear Supplier No. MM70186849
  • ONLINE EXCLUSIVE PRICE
    136.50 €
    86.10 € / 1 pairs
    Save 50.40 €
    36% Off
Estimated Shipment: 04-02-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
All purpose safety boot. The outsole repels acid and oil like an umbrella repels water. Strong ankle supporting upper fits the foot snugly and comfortably.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Duo Heat Resistant Outsole
  • Hydro-Tec™ Sanitized Silver for drier, fresher feet
  • Sanitized
Specifications
Design BAS XD
Material Leather
Size 49