Learn More
SCAT Base for space saving canister
Shop All SCAT Europe Products
Click to view available options
:
Material: Stainless steel, Typ Canister Holder
Description
- Made of stainless steel
- Base for 2 space saving containers
SCAT space-saving canisters offer a clever alternative to conventional SCAT canisters by requiring only a fraction of the floor space for storage, without compromising on capacity
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Color | Translucent White |
| Description | Base for 2 space saving containers |
| Dimensions (L x W x H) | 200 x 141 x 130 mm |
| Material | Stainless Steel |
| Product Type | Jerry Can Support Stand |
| Sterility | Non-sterile |