missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Basic Fuchsin, 88%, pure

Product Code. 10455661 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10455661 100 g Glass bottle 100g
10137610 500 g Glass bottle 500g
10369053 2.5 kg Plastic drum 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10455661 Supplier Thermo Scientific Chemicals Supplier No. 419141000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 632-99-5 | C20H20ClN3 | 337.85 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-99-5
Molecular Formula C20H20ClN3
Molecular Weight (g/mol) 337.85
MDL Number MFCD00012569
InChI Key AXDJCCTWPBKUKL-UHFFFAOYSA-N
Synonym Basic Violet 14, hydrochloride, C.I. 42510, Rosaniline chloride
PubChem CID 12447
IUPAC Name 4-[(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline;hydrochloride
SMILES [H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1

Specifications

Absorbance 1800 (at lambda min.)
Melting Point 250.0°C
Absorption Ratio 1.1 to 1.4
CAS Min % 83.0
CAS Max % 100.0
Dye Content 83% min. (on dry substance)
Infrared Spectrum Authentic
Lambda Max 549 to 555nm (in 50:50 IMS:water)
Packaging Glass bottle
Assay Percent Range 88%
Beilstein 13, 765
Merck Index 15, 5715
Color Green
Solubility Information Solubility in water: 4g/L (25°C). Other solubilities: soluble in alcohol and acids, practically insoluble in ether, soluble 30g/L in ethanol (25°C)
Quantity 100 g
Flash Point 200°C
Physical Form Fine Powder
Formula Weight 337.84
Percent Purity 88%
Grade Pure
Chemical Name or Material Basic Fuchsin, 88%
Show More Show Less
compliance-icons
Product Identifier
  • Basic Fuchsin
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Magenta

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.