missing translation for 'onlineSavingsMsg'
Learn More

Brannan™ Batch Sampling Blood Bank Thermometer with Calibration Certificate

Product Code. 17852978
Change view
Click to view available options
Calibrated:
3 Point Traceable
3 UKAS Traceable
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Calibrated unitSize
17852978 3 Point Traceable Each
17823108 3 UKAS Traceable Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17852978 Supplier Brannan™ Supplier No. 22/100/2 + 49/101/0
Only null left
Add to basket
Add to basket

Precision test batch sampling blood bank thermometers sealed into a 30 mL polypropylene bottle containing a heat transfer medium, ideally suited to monitor the temperature of samples in a controlled storage. Complete with traceable calibration.

  • Non-hazardous LO-tox™ filling
  • Bottle should be filled with monpropylene glycol or other heat transfer medium before use
  • Temperature measurement unaffected by short term fluctuations
  • Temperature range: -5°C to +20°C
  • Supplied with calibration and statement of accuracy
  • Made in UK

Specifications

Bottle Fill Customer to fill with monopropylene glycol or other heat transfer medium before use
Calibration Points 0°C, 10°C, 20°C
Certifications/Compliance UK National Standards ISO 9001
For Use With (Application) Monitor temperature of samples in a controlled storage
Height (English) 5.71 in.
Height (Metric) 14.5 cm
Includes Polypropylene bottle and glass thermometer
Length (Metric) 14.5 cm
Material Glass
Thermometer Fill No
Weight (English) 1.9577 oz.
Weight (Metric) 55.5 g
Width (English) 0.14 in.
Max. Temperature (English) 68°F
Max. Temperature (Metric) +20°C
Min. Temperature (English) 23°F
Min. Temperature (Metric) -5°C
Temperature Range (English) 23°F to 68°F
Temperature Range (Metric) -5°C to +20°C
Calibrated 3 Point Traceable
For Use With (Equipment) Blood bank refrigeration
Battery Type None
Accuracy ± 0.7°C
Subdivision 0.5°C
Display None
Width (Metric) 3.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.