missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brannan™ Batch Sampling Blood Bank Thermometer with Calibration Certificate
Description
- Non-hazardous LO-tox™ filling
- Bottle should be filled with monpropylene glycol or other heat transfer medium before use
- Temperature measurement unaffected by short term fluctuations
- Temperature range: -5°C to +20°C
- Supplied with calibration and statement of accuracy
- Made in UK
Specifications
Specifications
| Bottle Fill | Customer to fill with monopropylene glycol or other heat transfer medium before use |
| Calibration Points | 0°C, 10°C, 20°C |
| Certifications/Compliance | UKAS |
| For Use With (Application) | Monitor temperature of samples in a controlled storage |
| Height (English) | 5.71 in. |
| Height (Metric) | 14.5 cm |
| Includes | Polypropylene bottle and glass thermometer |
| Length (Metric) | 14.5 cm |
| Material | Glass |
| Thermometer Fill | No |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?