missing translation for 'onlineSavingsMsg'
Learn More

Bathocuproin sulfonate disodium salt hydrate, 97%

Product Code. 11354599
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354599 250 mg 250mg
11445117 1 g 1g
11455117 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11354599 Supplier Thermo Scientific Alfa Aesar Supplier No. B22550.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52698-84-7 | C26H18N2Na2O6S2 | 564.54 g/mol

Used as a stain for proteins in polyacrylamide gels; for determination of iron and copper; and uric acid by automated proceduresBathocuproin sulfonate disodium salt hydrate is a mixture of isomers suitable for the determination of iron and copper. It is also used as a stain for proteins in polyacrylamide gels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a stain for proteins in polyacrylamide gels; for determination of iron and copper; and uric acid by automated proceduresBathocuproin sulfonate disodium salt hydrate is a mixture of isomers suitable for the determination of iron and copper. It is also used as a stain for proteins in polyacrylamide gels.

Solubility
Miscible with water.

Notes
Incompatible with sulfuric acid, sodium metal, potassium metal and calcium carbide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52698-84-7
Molecular Formula C26H18N2Na2O6S2
Molecular Weight (g/mol) 564.54
MDL Number MFCD00149974
InChI Key RNGKZLRAVYPLJC-UHFFFAOYSA-L
Synonym disodium bathocuproinedisulfonate, sodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, bathocuproinedisulfonic acid sodium salt, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate, 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline disulfonate disodium salt, dipotassium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate, disodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate
PubChem CID 15678335
SMILES [Na+].[Na+].CC1=NC2=C(C=CC3=C2N=C(C)C(=C3C2=CC=CC=C2)S([O-])(=O)=O)C(C2=CC=CC=C2)=C1S([O-])(=O)=O

Specifications

Melting Point >300°C
Quantity 250 mg
Beilstein 9033637
Solubility Information Miscible with water.
IUPAC Name disodium;2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate
Formula Weight 564.55 (Anhydrous)
Percent Purity 97%
Chemical Name or Material Bathocuproin sulfonate disodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.