missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Bathophenanthroline

Product Code. 16462399
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16462399 1 g 1g
16402316 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462399 Supplier MedChemExpress Supplier No. HYD0006
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Bathophenanthroline is an agent used for the measurement of iron concentrations in aqueous , serum and urine samples by colorimetry.

Specifications

Quantity 1 g
Storage Requirements 4°C, protect from light, stored under nitrogen ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light, stored under nitrogen)
Solubility DMSO : 2.78 mg/mL (8.36 mM; Need ultrasonic) ∣ H2O : < 0.1 mg/mL (insoluble)
Shipping Condition Room temperature
CAS 1662-01-7
Molecular Weight (g/mol) 332.4 g/mol
Target HBV
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.