missing translation for 'onlineSavingsMsg'
Learn More

Bathophenanthroline, 98+%

Product Code. 11472397
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472397 25 g 25g
11452397 1 g 1g
11462397 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11472397 Supplier Thermo Scientific Alfa Aesar Supplier No. A14258.14
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1662-01-7 | C24H16N2 | 332.406 g/mol

Bathophenanthroline is widely used for determination of iron in serum and urine by colorimtetry. It acts as buffer layer to improve the efficiency of organic photovoltaic cells. It is very useful as a scavenger reagent, for the removal of traces of iron and copper from reagent solutions, which is used in the trace metal determination. Iron bathophenanthroline complex is involved as a redox mediating agent for imaging surface immobilized deoxyribonucleic acid (DNA) by using Scanning Electrochemical Microscopy (SECM).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bathophenanthroline is widely used for determination of iron in serum and urine by colorimtetry. It acts as buffer layer to improve the efficiency of organic photovoltaic cells. It is very useful as a scavenger reagent, for the removal of traces of iron and copper from reagent solutions, which is used in the trace metal determination. Iron bathophenanthroline complex is involved as a redox mediating agent for imaging surface immobilized deoxyribonucleic acid (DNA) by using Scanning Electrochemical Microscopy (SECM).

Solubility
Soluble in organic solvents. Slightly soluble in acidic aqueous solutions. Insoluble in neutral or alkaline solutions.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1662-01-7
Molecular Formula C24H16N2
Molecular Weight (g/mol) 332.406
MDL Number MFCD00004976
InChI Key DHDHJYNTEFLIHY-UHFFFAOYSA-N
Synonym bathophenanthroline, 1,10-phenanthroline, 4,7-diphenyl, bphen, 1,10-bathophenanthroline, bathophenanthrolin, bathophenanthrolin german, unii-4a2b091f0g, 4,7-diphenyl-1,10-diazaphenanthrene, gnf-pf-4554, 4,7-diphenylpyridino 3,2-h quinoline
PubChem CID 72812
ChEBI CHEBI:77995
IUPAC Name 4,7-diphenyl-1,10-phenanthroline
SMILES C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5

Specifications

Melting Point 218°C to 222°C
Quantity 25 g
Beilstein 261048
Solubility Information Soluble in organic solvents. Slightly soluble in acidic aqueous solutions. Insoluble in neutral or alkaline solutions.
Formula Weight 332.41
Percent Purity ≥98%
Chemical Name or Material Bathophenanthroline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.