missing translation for 'onlineSavingsMsg'
Learn More

Bathophenanthroline, MP Biomedicals™

Product Code. 11414612 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414612 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11414612 Supplier MP Biomedicals Supplier No. 0219015701
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 1662-01-7
Molecular Formula C24H16N2
Quantity 1 g
Synonym bathophenanthroline, 1,10-phenanthroline, 4,7-diphenyl, bphen, 1,10-bathophenanthroline, bathophenanthrolin, bathophenanthrolin german, unii-4a2b091f0g, 4,7-diphenyl-1,10-diazaphenanthrene, gnf-pf-4554, 4,7-diphenylpyridino 3,2-h quinoline
InChI Key DHDHJYNTEFLIHY-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5
IUPAC Name 4,7-diphenyl-1,10-phenanthroline
Molecular Weight (g/mol) 332.406
PubChem CID 72812
ChEBI CHEBI:77995
Formula Weight 332.41
Physical Form Solid
Chemical Name or Material Bathophenanthroline
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.