missing translation for 'onlineSavingsMsg'
Learn More

Bathophenanthrolinedisulfonic acid, disodium salt hydrate, 97%

Product Code. 10428730 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10428730 1 g Glass bottle 1g
10584454 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10428730 Supplier Thermo Scientific Acros Supplier No. 164050010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 52746-49-3 | C24H14N2Na2O6S2 | 536.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52746-49-3
Molecular Formula C24H14N2Na2O6S2
Molecular Weight (g/mol) 536.48
MDL Number MFCD00149310
InChI Key PCNDSIWXTYFWIA-UHFFFAOYSA-L
Synonym 4,7-Diphenyl-1,10-phenanthrolinedisulfonic acid, disodium salt hydrate
PubChem CID 54669730
IUPAC Name disodium;4-[7-(4-sulfonatophenyl)-1,10-phenanthrolin-4-yl]benzenesulfonate;hydrate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)C1=CC=NC2=C1C=CC1=C2N=CC=C1C1=CC=C(C=C1)S([O-])(=O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color Beige-Pink
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Formula Weight 536.5
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Bathophenanthrolinedisulfonic acid, disodium salt hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.