missing translation for 'onlineSavingsMsg'
Learn More

Asecos™ BATTERY STORE Storage Cabinet ION-CLASSIC-90

Product Code. 16359426 Shop All asecos Products
Change view
Click to view available options
No. of Doors:
1
2
No. of Shelves:
3
4
5
6
Unit Size:
Each
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Doors No. of Shelves unitSize
16359426 1 3 Each
16369426 1 4 Each
16379426 1 5 Each
16389426 1 6 Each
16379406 2 3 Each
16389406 2 4 Each
16399406 2 5 Each
16309416 2 6 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy

For indoor use only

Product Code. 16359426 Supplier Asecos™ Supplier No. 3806704738086

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For indoor use only

Safe passive storage of lithium-ion batteries

  • Type 90 Safety storage cabinet
  • Tall cabinet with self-closing wing doors
  • With cylinder locking and locking state indicator
  • Integrated adjusting aids to compensate uneven floor
  • Integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
  • Safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Integrated transport base with removable, optional base cover
  • Fire resistance 90 minutes
  • Type tested in accordance with EN 14470-1
  • CE compliant

Specifications

Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, CE compliant
Dimensions (D x W x H) Interior 522 x 450 x 1647 mm
Dimensions (D x W x H) 615 x 599 x 2224 mm
Door Style Wing
For Use With (Application) Safe passive storage of lithium-ion batteries
Includes Bottom collecting sump (sheet steel powder-coated)
Model IO90.195.060.K1.WDC
No. of Doors 1
No. of Shelves 3
Shelf Type Grid
Surface Coating Powder-coated textured
Color Anthracite Gray, Gentian Blue
Material Sheet Steel
Product Type Storage cabinet
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.