missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Bax inhibitor peptide V5

Product Code. 16433976
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16433976 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16433976 Supplier MedChemExpress Supplier No. HYP0081

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bax inhibitor peptide V5 (BIP-V5) is a Bax-mediated apoptosis inhibitor, used for cancer treatment.

Specifications

Purity 98.12%
CAS 579492-81-2
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 586.79 g/mol
Product Type Reference compound
Quantity 25 mg
Shipping Condition Room temperature
Solubility DMSO : ≥ 100 mg/mL (170.42 mM) ∣H2O : 100 mg/mL (170.42 mM∣Need ultrasonic)
Synonym BIP-V5∣ BAX Inhibiting Peptide V5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.