missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BAY 299

Description
BAY 299 is a potent and selective BRD1 and TAF1 inhibitor (IC50 values are 6-67 and 8-13 nM, respectively). Displays selectivity over other bromodomains (>30-fold over other members of the BRPF family; BRD9 and ATAD2; >300-fold over BRD4). Displays BRD1 and TAF1 inhibition in a NanoBRET cell assay. Inhibits binding of BRD1 and TAF1 to histone H4 (IC50 values are 575 nM and 0.9 μM, respectively) and histone H3.3 (IC50 values are 825 nM and 1.4 μM, respectively).
Specifications
Specifications
| Chemical Name or Material | 6-(3-Hydroxypropyl)-2-(1,3,6-trimethyl-2-oxo-2,3-dihydro-1H-benzimidazol-5-yl)-1H-benzo[de]isoquinoline-1,3(2H)-dione |
| CAS | 2080306-23-4 |
| Assay Percent Range | >98% |
| Quantity | 25 mg |
| Molecular Formula | C25H23N3O4 |
| Formula Weight | Observed MW: 429.47 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?