missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BAY 299
GREENER_CHOICE

Product Code. p-7116090
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
50mg
5mg
This item is not returnable. View return policy

Product Code. 18704039

Brand: Tocris Bioscience™ 5970/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective BRD1 and TAF1 inhibitor

BAY 299 is a potent and selective BRD1 and TAF1 inhibitor (IC50 values are 6-67 and 8-13 nM, respectively). Displays selectivity over other bromodomains (>30-fold over other members of the BRPF family; BRD9 and ATAD2; >300-fold over BRD4). Displays BRD1 and TAF1 inhibition in a NanoBRET cell assay. Inhibits binding of BRD1 and TAF1 to histone H4 (IC50 values are 575 nM and 0.9 μM, respectively) and histone H3.3 (IC50 values are 825 nM and 1.4 μM, respectively).
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-(3-Hydroxypropyl)-2-(1,3,6-trimethyl-2-oxo-2,3-dihydro-1H-benzimidazol-5-yl)-1H-benzo[de]isoquinoline-1,3(2H)-dione
CAS 2080306-23-4
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C25H23N3O4
Formula Weight Observed MW: 429.47
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.