missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ BAY 41-2272

Product Code. 18065322
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18065322 10 mg 10mg
18045832 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18065322 Supplier R&D Systems™ Supplier No. 4430/10
  • ONLINE EXCLUSIVE PRICE
    345.81 €
    309.75 € / 10mg
    Save 36.06 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Soluble guanylyl cyclase (sGC) activator

Activator of soluble guanylyl cyclase (sGC); acts at a nitric oxide (NO)-independent regulatory site in the sGC α1 subunit. Inhibits platelet aggregation (IC50 = 36 nM) and phenylephrine-induced contractions of rabbit aorta (IC50 = 0.30 μM). Also reduces vascular smooth muscle growth through cAMP- and cGMP-dependent PKA and PKG pathways.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 256376-24-6
Molecular Formula C20H17FN6

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 360.39
Purity >98%
Molecular Weight (g/mol) 360.40
Chemical Name or Material BAY 41-2272
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.