missing translation for 'onlineSavingsMsg'
Learn More

Tocris™ BAY 87-2243

Product Code. 18778836
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18778836

Brand: Tocris™ 6980/5

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mitochondrial complex I inhibitor

BAY 87-2243 is a mitochondrial complex I inhibitor. Exhibits no effect on mitochondrial complex III. Induces mitochondrial permeability transition pore (mPTP) opening, stimulates autophagosome formation and mitophagy, and increases ROS levels. Activates necroptotic and ferroptotic cell death. Also inhibits hypoxia-induced gene activation. Induces cell death in melanoma cell lines in vitro and exhibits antitumor activity in a mouse NSCLC xenograft model in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Cyclopropyl-4-[4-[[5-methyl-3-[3-[4-(trifluoromethoxy)phenyl]-1,2,4-oxadiazol-5-yl]-1H-pyrazol-1-yl]methyl]-2-pyridinyl]piperazine
CAS 1227158-85-1
Quantity 5 mg
Molecular Formula C26H26F3N7O2
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.