missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris™ BAY 87-2243
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
BAY 87-2243 is a mitochondrial complex I inhibitor. Exhibits no effect on mitochondrial complex III. Induces mitochondrial permeability transition pore (mPTP) opening, stimulates autophagosome formation and mitophagy, and increases ROS levels. Activates necroptotic and ferroptotic cell death. Also inhibits hypoxia-induced gene activation. Induces cell death in melanoma cell lines in vitro and exhibits antitumor activity in a mouse NSCLC xenograft model in vivo.
Specifications
Specifications
| Chemical Name or Material | 1-Cyclopropyl-4-[4-[[5-methyl-3-[3-[4-(trifluoromethoxy)phenyl]-1,2,4-oxadiazol-5-yl]-1H-pyrazol-1-yl]methyl]-2-pyridinyl]piperazine |
| CAS | 1227158-85-1 |
| Quantity | 25 mg |
| Molecular Formula | C26H26F3N7O2 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction