missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ (±)-Bay K 8644

Product Code. 18765936
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18765936 10 mg 10mg
18075012 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18765936 Supplier R&D Systems™ Supplier No. 1544/10
  • ONLINE EXCLUSIVE PRICE
    312.05 €
    278.25 € / 10mg
    Save 33.80 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. CaV1.x activator

L-type Ca2+ channel activator (EC50 = 17.3 nM). Has positive inotropic, vasoconstrictive and behavioral effects in vivo. In combination with BIX-01294 (Cat. No. 3364), helps generate induced pluripotent stem cells (iPSCs) from mouse embryonic fibroblasts (MEFs). Inhibits autophagy.

(S)-(-)-Enantiomer and (R)-(+)-Enantiomer also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71145-03-4
Molecular Formula C16H15F3N2O4

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 356.3
Purity >99%
Molecular Weight (g/mol) 356.301
Chemical Name or Material (±)-Bay K 8644
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.