missing translation for 'onlineSavingsMsg'
Learn More

Bazedoxifene Bis-Beta-D-Glucuronide Disodium Salt, TRC

Product Code. 30337783
Change view
Click to view available options
Quantity:
0.25 mg
Unit Size:
0.25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30337783 0.25 mg 0.25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337783 Supplier Toronto Research Chemicals Supplier No. TRCB1292650.25MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bazedoxifene Bis-beta-D-Glucuronide Disodium Salt
Molecular Formula C42 H48 N2 Na2 O15
Quantity 0.25 mg
Synonym 4-[5-(beta-D-Glucopyranuronosyloxy)-1-[[4-[2-(hexahydro-1H-azepin-1-yl)ethoxy]phenyl]methyl]-3-methyl-1H-indol-2-yl]phenyl beta-D-Glucopyranosiduronic Acid
InChI Formula InChI=1S/C42H50N2O15.2Na/c1-22-28-20-27(57-42-36(50)32(46)34(48)38(59-42)40(53)54)14-15-29(28)44(21-23-6-10-25(11-7-23)55-19-18-43-16-4-2-3-5-17-43)30(22)24-8-12-26(13-9-24)56-41-35(49)31(45)33(47)37(58-41)39(51)52;;/h6-15,20,31-38,41-42,45-50H,2-5,16-19,21H2,1H3,(H,51,52)(H,53,54);;/q;2*+1/p-2/t31-,32-,33-,34-,35+,36+,37-,38-,41+,42+;;/m0../s1
SMILES Cc1c(c2ccc(O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O[Na])cc2)n(Cc4ccc(OCCN5CCCCCC5)cc4)c6ccc(O[C@@H]7O[C@@H]([C@@H](O)[C@H](O)[C@H]7O)C(=O)O[Na])cc16
IUPAC Name [(2S,3S,4S,5R,6S)-6-[4-[1-[[4-[2-(azepan-1-yl)ethoxy]phenyl]methyl]-3-methyl-5-[(2S,3R,4S,5S,6S)-3,4,5-trihydroxy-6-sodiooxycarbonyloxan-2-yl]oxyindol-2-yl]phenoxy]-3,4,5-trihydroxyoxane-2-carbonyl]oxysodium
Molecular Weight (g/mol) 866.814
Formula Weight 866.285
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.