missing translation for 'onlineSavingsMsg'
Learn More

BD BB515 Streptavidin

Product Code. 15893649
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15893649 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15893649 Supplier BD Supplier No. 564453

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Affinity purified

Host Species:
Species Reactivity: Mouse

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Streptavidin
Conjugate BB515
Cross Reactivity Mouse
Quantity 0.1 mg
Form Affinity-purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.