Learn More
BD BBL™ Crystal™ ID Systems Accessories
Product Code. 11778049
Change view
Click to view available options
Product Type:
Anaerobic Nh inoculum
Calibration Standard
Microbiology Media
Quantity:
10 Pack
Each
Unit Size:
Each
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Quantity Unit Size
11778049 Calibration Standard Each Each
11768912 Anaerobic Nh inoculum 10 Pack Pack of 10
11738912 Microbiology Media 10 Pack Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11778049 Supplier BD Supplier No. 245015
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
BBL Crystal MIND Software
  • Interprets the test results and provides specimen identification
  • Minimum software requirements are an MS-DOS™ 5.0 or higher compatible computer; Intel™ 486 microprocessor with at least 16MB RAM Software is compatible with Microsoft™ Windows™ (3.11, 95, 98, 2000, XP) Computer must have at least Microsoft ODBC Version 2.00 and a 3.5 in. floppy drive
Codebooks Include Enteric/Nonfermenter, Anaerobe, Neisseria/Haemophilus, Gram-Positive, Rapid Gram-Positive) Supplied in individual hard-copy binder BBL Crystal Panel Viewer
  • For facilitated viewing and recording of chromogenic and fluorogenic reactions
BBL CrystalSpec™ Standards†
  • Set of two calibration standards for the CrystalSpec Nephelometer, one Calibration Blank and one Calibration Standard
†Drop shipment is required. All shipments FOB from Cockeysville, MD. Freight charges will be prepaid and billed as a separate item on the invoice.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Calibration Standard
For Use With (Equipment) CrystalSpec™ nephelometer
Includes Set of two calibration standard, one calibration blank and one calibration standard
Quantity Each