missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ CultureSwab EZ™ and CultureSwab EZ II™

Product Code. 14871121
Change view
Click to view available options
Swab Configuration:
Single Applicator
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Swab Configuration Unit Size
14871121 Single Applicator Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 14871121 Supplier BD Supplier No. 220144
  • ONLINE EXCLUSIVE PRICE
    279.27 €
    226.17 € / Pack of 100
    Save 53.10 €
    19% Off
 Restricted Item
Estimated Shipment: 11-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Polyurethane sponge swab on a plastic shaft secured to a cap, which is inserted into supplied plastic transport tube

A patented media-free collection system designed to collect aerobic specimens and maintain viability during transport

  • No specimen dilution with this dry system and no ampule to crush
  • Transport system eliminates the organism overgrowth and nonviables typically present in agar-based systems
  • Aerobic microorganisms remain viable for approximately 48 hours
  • Features rounded tube bottom for easier handling, and water-resistant film packing with tamper-evident seal
  • BD CultureSwab EZ II is similar to BD CultureSwab EZ except formatted as a dual swab system, allowing for simultaneous gram staining and other direct antigen testing

TRUSTED_SUSTAINABILITY

Specifications

Product Type Culture Swab™ EZ System
Sterility Sterile
Swab Configuration Single Applicator
For Use With (Application) For collecting and transporting aerobic microbiological specimens
Includes Polyurethane single sponge swab, Plastic shaft, cap, inserted into Plastic transport tube
Quantity 100/Pk.
Shaft Material Plastic
Storage Requirements 15°C to 30°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.