missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Phosphate Buffered Saline

Product Code. 11742774
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging Type:
Bottle
Vial
Unit Size:
100g
60g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type unitSize
11742774 100 g Bottle 100g
11752774 10 g Vial 60g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11742774 Supplier BD Supplier No. 223142

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorescent Antibody reagents

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage <30°C
Additive Phosphate Buffered Saline
Form Dehydrated
For Use With (Application) Fluorescent Antibody reagents
Packaging Type Bottle
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.