Learn More
BD BBL™ Prepared Lowenstein-Jensen Media
Used for cultivation of Mycobacterium tuberculosis and other mycobacterial species
Brand:  BD 220909
Additional Details : Weight : 3.70800kg
 View more versions of this product
Product Code. 11938536
  • 610.00 € / Pack of 100
Estimated Shipment: 10-06-2024
to see stock.
Add to basket
Description
Description
  • Inspissated, egg-based media
  • Available as Mycoflask™ bottles, prepared slants and bottles
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
Specifications
CE, BS12, CMPH2, EP, MCM9, SMWW
2°C to 8°C
United States
Monopotassium Phosphate 2.5g/L, Magnesium Sulfate 0.24g/L, Sodium Citrate 0.6g/L, L-Asparagine 3.6g/L, Potato Flour 30g/L, Malachitegreen 0.4g/L, Glycerol 12mL, Whole Egg 1000mL
Slants (A tubes)
Beige
Liquid
Lowenstein-Jensen Media
Cultivation of Mycobacterium tuberculosis and other mycobacterial species
100/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For in vitro diagnostic use.