missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ BCR™ Single-Knit Nylon Full-Finger Cleanroom Glove Liners

Product Code. 17154824 Shop All Berkshire Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
200 pairs
201 pairs
203 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17154824 Small 200 pairs
17144824 Medium 201 pairs
17134824 Large -
17164824 X-Large 203 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17154824 Supplier Berkshire™ Supplier No. BGL7.200SB
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Single-knit nylon construction is thin and comfortable, while the full-finger design provides total protection for those with severe glove sensitivities.

  • Reduces perspiration and contact dermatitis commonly associated with wearing latex, urethane, PVC, or nitrile gloves
  • Full-finger design provides total protection for those with severe glove sensitivities
  • Can be worn alone as inspection gloves to prevent fingerprints on critical surfaces
  • Single-knit nylon construction is thin and comfortable
  • Ambidextrous design can be used on either hand
  • Gloves can be repeatedly laundered for reuse
  • Ideal for wet or temperature related processing
  • Suitable for use in ISO Class Cleanrooms

Specifications

Construction Single Knit
Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Material Nylon
Size Small
Finger Style Full
For Use With (Application) Wet/Temperature-Related Processing, Preventing Contact Dermetitis and Perspiration
Glove Cuff Style Knit
Hem Color Green
Series BGL7
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.