missing translation for 'onlineSavingsMsg'
Learn More

Berkshire™ BCR™ Nylon Glove Liners, Full-Finger

Product Code. 17164824 Shop All Berkshire Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
200 pairs
201 pairs
203 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17164824 X-Large 203 pairs
17154824 Small 200 pairs
17144824 Medium 201 pairs
17134824 Large -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17164824 Supplier Berkshire™ Supplier No. BGL7.200XLB
  • ONLINE EXCLUSIVE PRICE
    590.10 €
    438.93 € / 200 pairs
    Save 151.17 €
    25% Off
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nylon full-finger glove liners protect hands and improve operator comfort. Made from 100% knitted nylon, these reusable, seamless liners provide full-finger coverage, reduce perspiration, and support extended wear under cleanroom gloves.

  • Enhance operator comfort.
  • Greatly reduce perspiration and contact dermatitis associated with wearing latex, urethane, PVC, nitrile and other chemical handling gloves.
  • Liners can be repeatedly laundered and reused.
  • 100% knitted nylon, single-knit is thinner and comfortable.
  • Nylon liners are heavier and more durable and can withstand more laundry washes.
  • Full-finger design allows total protection for the most sensitive glove users.
  • Reusable, ambidextrous, seamless construction.
  • For use in laboratories and manufacturing environments.
  • Can be used alone as inspection gloves to prevent fingerprints on circuit boards, glassware, lenses, optics and other critical surface.
  • Nylon provides a thin barrier for wet and temperature related processing.
  • Suitable in any ISO class cleanroom when worn under a nitrile, latex, vinyl, neoprene or other type glove.

Specifications

Construction Single Knit
Coating Material Uncoated
Color White
Disposable Reusable
Hand Ambidextrous
Laboratory/Cleanroom Use Yes
Latex Free Yes
Material Nylon
Product Type Full Glove Liners
Size X-Large
Sterility Non-sterile
Style Full Finger Nylon
Finger Style Full
For Use With (Application) Laboratories, Manufacturing, Underglove, ISO Class Cleanroom, Wet/Temp Processing
Gender Unisex
Glove Cuff Color Red
Glove Cuff Style Knit
Hem Color Red
Packaging Type Bulk
Series BCR Glove Liners
Product Line BCR Cleanroom Glove Liners
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.