missing translation for 'onlineSavingsMsg'
Learn More

BD Acridine Orange Stain

Product Code. p-103472
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11738213 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11738213 Supplier BD Supplier No. 212536

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For detecting microorganisms in direct smears by fluorescence

  • Especially beneficial in screening normally sterile specimens where few organisms may be present
  • Bacteria stain bright orange; human cells, tissue debris and proteinaceous material stain pale green to yellow

TRUSTED_SUSTAINABILITY

Specifications

Type Acridine Orange Stain
Quantity 250 mL
Certifications/Compliance CE
For Use With (Application) Recommended for use in the fluorescent microscopic detection of microorganisms in direct smears prepared from clinical and non clinical materials
Stain Type Acridine Orange
compliance-icons
Product Identifier
  • Acridine Orange Stain
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P321-Specific treatment (see on this label).
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: hydrochloric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.