missing translation for 'onlineSavingsMsg'
Learn More

BD AEC Substrate Kit

Product Code. 15826478
Change view
Click to view available options
Chromagen Quantity:
1.5 mL
3 mL
Includes:
AEC buffer and AEC chromogen
Plates, AEC buffer, chromogen
Unit Size:
1 set
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Chromagen Quantity Includes unitSize
15826478 1.5 mL AEC buffer and AEC chromogen 1 set
15836738 3 mL Plates, AEC buffer, chromogen Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Contains Dimethylsulfoxide, Methanol, Urea Peroxide, 3-Amino-9-Ethylcarbazole, and N, N,Dimethylformamide which are considered as harmful.
Product Code. 15826478 Supplier BD Supplier No. 551015

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Contains Dimethylsulfoxide, Methanol, Urea Peroxide, 3-Amino-9-Ethylcarbazole, and N, N,Dimethylformamide which are considered as harmful.

Includes

AEC Buffer and AEC Chromogen

Substrate for peroxidase and is a widely used chromogen for immunohistochemical staining and immunoblotting

Immunohistochemistry, ELISPOT

TRUSTED_SUSTAINABILITY

Specifications

Chromagen Quantity 1.5 mL
Chromagen Type AEC
For Use With (Application) Immunohistochemical staining and immunoblotting, ELISPOT
Substrate Type AEC Substrate
Includes AEC buffer and AEC chromogen
pH 7.2
Storage Requirements 4°C
compliance-icons compliance-icons compliance-icons
Product Identifier
  • AEC Substrate Kit
Signal Word
  • Danger||Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: methanol, N,N-dimethyl formamide, 9-ethylcarbazol-3-ylamine

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.