missing translation for 'onlineSavingsMsg'
Learn More

BD AKP Streptavidin

Product Code. 15886468
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15886468 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15886468 Supplier BD Supplier No. 551008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pre-diluted, ready-to-serve, convenient to use enzyme which provides optimal signal amplification and detection of an antigen

  • Storage buffer: Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.

Immunohistochemistry

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Quantity 50 mL
Content And Storage 4°C
Form Liquid
For Use With (Application) Immunohistochemistry

Research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.