missing translation for 'onlineSavingsMsg'
Learn More

BD Annexin V Binding Buffer

Product Code. 15878698
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15878698 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15878698 Supplier BD Supplier No. 556454
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Formulated for use in flow cytometric assays utilizing Annexin V

  • Composed of 0.2μm sterile filtered 0.1M Hepes (pH 7.4), 1.4M NaCl, and 25mM CaCl2 solution
  • 1X working solution should be made by diluting the 10X concentrate
  • Remaining 1X solution at the end of the experiment should be discarded
  • Format: Buffer

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 mL
Content And Storage Store at 4°C

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.