missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Annexin V, BUV395, Recombinant Protein
Click to view available options
:
100 Tests, Brilliant Ultraviolet 395
Specifications
Specifications
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide |
| Molecular Weight (g/mol) | 31-35kDa |
| Storage Requirements | 4°CXXXXX protect from prolonged exposure to light. Do not freeze. |
| Regulatory Status | RUO |
| Conjugate | BUV395 |
| Cross Reactivity | Human |
| Recombinant | Recombinant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction