missing translation for 'onlineSavingsMsg'
Learn More

BD Anti-mouse Ig HRP Detection kit

Product Code. 15896468 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15896468 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15896468 Supplier BD Supplier No. 551011
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Biotinylated goat anti-mouse Igs, Antibody Diluent Buffer, Streptavidin-Horseradish peroxidase (SAv-HRP), DAB Buffer, and DAB Chromogen

Designed for immunohistochemical staining of acetone-fixed frozen sections, formalin- or zinc-fixed paraffin sections, and cytospins/smears

  • Detects molecules within a tissue microenvironment
  • Protein expression can be studied in relation to tissue or cell compartments
  • Also changes due to either normal or pathologic stimuli can be evaluated
  • Detection of an antibody bound to antigen can be accomplished by direct or indirect methods
  • Size: 200 tests
  • Regulatory status: RUO

Immunohistochemistry

TRUSTED_SUSTAINABILITY

Specifications

Storage Requirements 4°C
For Use With (Application) Immunohistochemistry, Immunohistochemical Staining
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.