missing translation for 'onlineSavingsMsg'
Learn More

BD APC BrdU Flow Kit, 50 tests

Product Code. 10773807
Change view
Click to view available options
Quantity:
50 Tests
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10773807 50 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10773807 Supplier BD Supplier No. 552598
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Comes with 65μL of APC-conjugated anti-BrdU antibody

Provides reagents for immunofluorescent BrdU staining with a protocol, compatible with the use of additional fluorescent antibodies specific for other cellular molecules. Used with fluorescent anti-cytokine antibodies in analyses of cultured cells following in vitro mitogenic stimulation of quiescent lymphoid cell populations.

 

  • Consists of Flow cytometer with 488nm laser capable of detecting 7-AAD and a 633- to 640nm laser capable of detecting APC
  • Staining procedure includes the fixative paraformaldehyde

 

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance RUO
Content And Storage 4°C
For Use With (Application) For immunofluorescent BrdU staining, fluorescent Anti-cytokine antibodies in analyses of cultured cells
For Use With (Equipment) Flow cytometer
Includes One 65μL vial of APC-conjugated Anti-BrdU antibody, one 25mL bottle Cytofix/Cytoperm™ buffer, two 25mL bottles perm/Wash™ buffer (10X), one 10mL bottle Cytoperm™
No. of Tests 50
Quantity 50 Tests
Product Type APC BrdU Flow Kit
compliance-icons compliance-icons compliance-icons
Product Identifier
  • APC BrdU Flow Kit
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1A
  • Germ cell mutagenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: formaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.