missing translation for 'onlineSavingsMsg'
Learn More

BD B Lymphocyte Subset Antibody Cocktail

Product Code. 15879308 Shop All BD Biosciences Products
Change view
Click to view available options
Product Type:
B lymphocyte subset antibody cocktail
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15879308 B lymphocyte subset antibody cocktail Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15879308 Supplier BD Supplier No. 558331
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • Clones: B3B4, RA3-6B2, II/41
  • Formats: PE, PE-Cy 7, FITC
  • Isotype: Rat IgG2a, κ

Three-color reagent designed to identify major subsets of B lymphocytes

The Mouse B Lymphocyte Subset Antibody Cocktail is a three-color reagent designed to identify major subsets of B lymphocytes by direct immunofluorescent staining with flow cytometric analysis.

  • Three different fluorochromes label three different antibodies, permitting recognition of each of the three antigens on each cell in a sample
  • The three antibodies have been titrated and pre-diluted, mixed together and formulated for optimal staining performance
  • Levels of expression of the three antigens distinguish the major subpopulations of developing and peripheral B lymphocytes
  • Additional fluorochrome-labelled reagents may be combined with the Mouse B Lymphocyte Subset Antibody Cocktail to further characterize B-cell subpopulations

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Clone B3B4, RA3-6B2, II/41
Content And Storage Undiluted; 4°C Protect from prolonged exposure to light; Do not freeze
Product Type B lymphocyte subset antibody cocktail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.