missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Difco™ Casamino Acids, vitamin assay

Product Code. 16219861 Shop All Gibco Products
Change view
Click to view available options
Quantity:
100 g
500 g
For Use With (Application):
Vitamin assay
Grade:
Vitamin Assay
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity For Use With (Application) Grade Unit Size
16219861 100 g Vitamin assay Vitamin Assay 100g
16229861 500 g Vitamin assay Vitamin Assay 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16219861 Supplier Gibco™ Supplier No. 228820
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Excellent supplement for media formulations where nitrogen requirements are minimal, including diphtheria toxin production. Gibco™ Bacto™ Casamino Acids is an animal-origin, acid hydrolysis of casein having low sodium chloride and iron concentrations.

  • Prepared according to the method described by Mueller and Miller
  • Supplemented with growth factors, cystine, maltose and iron and inorganic salts
  • Successful in fermentation for Pneumococcal, Diptheria, Pertussis, Tetanus and Zoster vaccines
  • Support the growth of Bordetella, Corynebacterium, Pseudomonas, Haemophilus and Escherichia
  • Dehydrated format with Peptones / Hydrolysates
  • DNR (Latex) free
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, COMPF, SMWW, USDA, USP, ISO/DIS, 11014
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Additive Casamino Acid
Form Dehydrated Powder
Grade Vitamin Assay
Product Type Casamino Acids
For Use With (Application) Vitamin assay
Packaging Type Bottle
Product Line Difco
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.