missing translation for 'onlineSavingsMsg'
Learn More

BD Bacto™ Dehydrated Agar

Product Code. 10455513
Change view
Click to view available options
Quantity:
100 g
454 g
2000 g
Packaging Type:
Bottle
Container
Unit Size:
100g
2kg
454g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type unitSize
10455513 454 g Bottle 454g
11768223 100 g Bottle 100g
11758223 2000 g Container 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10455513 Supplier BD Supplier No. 214010
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Purified solidifying agent. BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter.

Purified solidifying agent. BD Difco™ Bacto™ Dehydrated Agar is purified to remove extraneous matter.

  • Extraneous matter, pigmented portions and salts are reduced to minimum
  • Fine, light colored granules
  • Convenient for weighing and handling

Determines motility and growth of anaerobes and microaerophiles

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Concentration 1.5%
Additive Agar
Form Dehydrated
For Use With (Application) Determination of motility and the growth of anaerobes and microaerophiles
Packaging Type Bottle
Quantity 454 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.