missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Bacto™ Yeast Extract
Click to view available options
Quantity:
500 g
2 kg
10 kg
50 kg
Unit Size:
10kg
2kg
500g
50kg
Description
- Bacto Yeast Extracts are concentrates of the water-soluble portion of autolyzed Saccharomyces cerevisiae cells, derived from primary grown baker's yeast
- These animal origin-free products are suitable for use as multi-functional nutritional supplements in mammalian cell culture, microbial fermentation, and insect cell culture applications
- Yeast extract provides essential water-soluble vitamins, amino acids, peptides, and carbohydrates to any medium formulation
- Bacto Yeast Extract is one of the most complete and versatile fermentation supplements available
- It is an important ingredient for the microbiological assay of vitamins
- Yeast extract is also of value in the assay of antibiotics
- B factor, a growth substance necessary for the production of rifampin in a Nocardia species, can be isolated from yeast extract
- Bacto Yeast Extract, Technical, was developed to provide products priced for the biotechnology market with acceptable clarity and growth-promoting characteristics
- Store in cool, dry conditions in well-sealed containers at 2°C to 25°C
Related yeast extract products include:
- Gibco Yeast Extract – Developed to provide a product for the biotechnology/pharmaceutical market with acceptable clarity and growth-promoting characteristics. Media formulations containing yeast extract are specified in standard methods for various applications.
- Difco Yeast Extract UF – Ultra-filtered and specifically designed for mammalian cell culture applications. With its low endotoxin level and high content of naturally occurring B vitamins, it can be used effectively to reduce the need for fetal bovine serum. It has an endotoxin level of less than or equal to 500 EU/g
- Difco Yeast Extract Low-Dusting (LD) – Designed to limit the amount of dust generated compared to other spray-dried yeast extracts
Specifications
Specifications
| Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
| Culture Type | Mammalian, Insect, Microbial |
| Form | Dehydrated |
| Product Type | Dehydrated Culture Media |
| For Use With (Application) | Formulation of culture media for the cultivation of heterotrophic microorganisms |
| Product Line | Bacto |
| Quantity | 50 kg |
| Source | Yeast, animal origin-free |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction