missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Mycophil™ Agar with Low pH
Click to view available options
:
Mycophil Agar, with Low pH; 500g
Specifications
Specifications
| Form | Dehydrated |
| Media Type | Mycophil Agar |
| Product Type | Agar |
| Formulation | Papaic digest of Soybean Meal 10g/L, Dextrose 10g/L, Agar 18g/L |
| For Use With (Application) | Isolation and enumeration of Yeast and molds |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction