missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Middlebrook OADC Enrichment

Product Code. 17434923
Change view
Click to view available options
Quantity:
6 x 100 mL
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17434923 6 x 100 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17434923 Supplier BD Supplier No. 212240
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used in qualitative procedures for isolation and cultivation of mycobacteria. BD Difco™ BBL™ Middlebrook OADC Enrichment is a dehydrated culture media.

  • 100 mL per bottle (6/sp)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2°C to 8°C
Additive Middlebrook OADC Enrichment
Form Viscous
For Use With (Application) Middlebrook 7H10 Agar
Packaging Type Bottle
Quantity 6 x 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.