missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ IsoVitaleX™ Enrichment

Product Code. 11798163
Change view
Click to view available options
Quantity:
5 x 10 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11798163 5 x 10 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11798163 Supplier BD Supplier No. 211876
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BD BBL™ IsoVitaleX™ Enrichment is a chemically defined supplement used as an additive to media for cultivation of nutritionally fastidious microorganisms.

  • GC Agar supplemented with hemoglobin and BBL IsoVitaleX Enrichment (in place of yeast concentrate) provides an improved medium for the isolation of microorganisms
  • Enrichment provides V factor (nicotinamide adenine dinucleotide, NAD) for Haemophilus species
  • Also provides vitamins, amino acids, coenzymes, dextrose, ferric ions and other factors which improve the growth of pathogenic Neisseria
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO/DIS, 11014
Content And Storage 2°C to 8°C
Additive IsoVitaleX™ Enrichment
Form Lyophilized
For Use With (Application) Dehydrated culture media
Packaging Type Vial
Quantity 5 x 10 mL
compliance-icons
Product Identifier
  • Isovitalex Enrichment
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P363-Wash contaminated clothing before reuse.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Hydrochloric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.