Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Disc Amikacin2, AN-30
Product Code. 17382381
Change view
Click to view available options
Concentration:
30 μg
Unit Size:
Pack of 10
Product Code. Concentration Unit Size
17382381 30 μg Pack of 10
1 options
Product Code. 17382381 Supplier BD Supplier No. 231597
Outlet Corner
Use promo code 28080
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Cost-effective method uses antimicrobial agents impregnated on high-quality paper
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents.
  • Employs Bauer-Kirby disc diffusion testing method
  • Use in self-tamping dispensers
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
  • Cartridges of 50 discs each are sold individually or in packs of 10
TRUSTED_SUSTAINABILITY
Specifications
Antimicrobial Agents Amikacin
Concentration 30 μg
Packaging Type Disc
Quantity 10/Pk.