missing translation for 'onlineSavingsMsg'
Learn More

BD Biotin Mouse CD4 T Lymphocyte Enrichment Set - DM

Product Code. 15869228
Change view
Click to view available options
Conjugate:
Biotin
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Conjugate Unit Size
15869228 Biotin Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15869228 Supplier BD Supplier No. 558131
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

CD8a, CD11b, CD45R/B220, CD49b, TER-119/Erythroid cells biotinylated antibody cocktail and BD IMag™ SAv particles

For negative selection of CD4 T lymphocytes from mouse spleen or lymph node

Components include Mouse CD4 T Lymphocyte Enrichment Cocktail and Streptavidin Particles Plus - DM

  • Reactivity: Mouse

Magnetic Labeling

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Biotin

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.