missing translation for 'onlineSavingsMsg'
Learn More

BD Biotin Mouse CD8 T Lymphocyte Enrichment Set - DM

Product Code. 15839418
Change view
Click to view available options
Quantity:
1 x 109 Leukocyte Cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15839418 1 x 109 Leukocyte Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15839418 Supplier BD Supplier No. 558471
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Enrichment cocktail (5mL): Anti-mouse CD4, clone GK1.5, Anti-mouse CD11b, clone M1/70, Anti-mouse CD45R/B220, clone RA3-6B2, Anti-mouse CD49b, clone HMα2, Anti-mouse TER-119/Erythroid Cells, clone TER-119 and SAv particles (5mL)

Negative selection of CD8 T lymphocytes from mouse spleen or lymph node

  • Monoclonal antibodies recognize antigens expressed on peripheral erythrocytes and leukocytes
  • Streptavidin covalently conjugated to surface of magnetic nanoparticles in Streptavidin Particles Plus - DM
  • Avoids activation of enriched CD8 T lymphocytes
  • Storage buffer comprises of aqueous buffered solution containing BSA and ≤0.09% sodium azide

Magnetic Labeling

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Biotin
Quantity 1 x 109 Leukocyte Cells

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.