missing translation for 'onlineSavingsMsg'
Learn More

BD Biotin Mouse Hematopoietic Progenitor (Stem) Enrichment Set - DM

Product Code. 15849398
Change view
Click to view available options
Quantity:
2 x 109 Cells
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15849398 2 x 109 Cells Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15849398 Supplier BD Supplier No. 558451
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Enrichment Cocktail (5mL): biotinylated monoclonal antibodies to mouse CD3ε (CD3 chain), CD11b (Integriα M chain), CD45R/B220, Ly-6G and Ly-6C (Gr-1) and TER-119/Erythroid Cells (Ly-76), SAv particles (5mL)

For immunomagnetic enrichment of hematopoietic progenitors

  • Optimized and pre-diluted to provide maximum efficiency
  • Reacts with cells from major hematopoietic cell lineages such as T lymphocytes, B lymphocytes, monocytes/macrophages, granulocytes and erythrocytes
  • Storage buffer comprises of aqueous buffered solution containing BSA and ≤0.09% sodium azide

Magnetic Labeling

TRUSTED_SUSTAINABILITY

Specifications

Conjugate Biotin
Quantity 2 x 109 Cells

Research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.