Learn More
BD BrdU In-Situ Detection Kit
Product Code. 15876388
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Product Code. Quantity Unit Size
15876388 1 Kit 1 set
1 options
Highly toxic, avoid contact with skin, eyes, and mucous membranes
Product Code. 15876388 Supplier BD Supplier No. 550803
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Highly toxic, avoid contact with skin, eyes, and mucous membranes
Includes
Fixation Buffer, Diluent Buffer, BD™ Retrievagen A Antigen Retrieval Solution, biotinylated anti-BrdU antibody, ready-to-use Streptavidin-Horseradish peroxidase (SAv-HRP) Solution, DAB Buffer, DAB Chromogen, and Control Slides
Requires
BrdU for in vitro or in vivo labeling, H2O2 (30%) solution for blocking endogenous peroxidase, phosphate buffer saline (PBS)
Determines proliferation rates and cell cycle kinetics in normal and cancerous tissues
Cellular incorporation of BrdU during S phase can be readily detected by anti-BrdU-specific antibodies following membrane permeabilization. Subsequent quantitation of cells positive for BrdU is possible using routine methods for cell analysis
  • Measures labeling indices of different cell populations
  • Improved specific staining with minimal background
  • Designed antigen retrieval solution unmask antigenic sites and preserve tissue morphology
  • Enables study of proliferation state of phenotypically defined cells within the micro-environment of tissues
  • Reaction: 50 slides
Flow Cytometry, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Research use only