missing translation for 'onlineSavingsMsg'
Learn More

BD Caspase-9 Inhibitor, Z-LEHD-FMK

Product Code. 15856258 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15856258 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15856258 Supplier BD Supplier No. 550381
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Irreversible and cell permeable inhibitor of caspase-9

Members of the caspase family play key roles in inflammation and mammalian apoptosis. The Z-LEHD-FMK peptide is O-methylated in the P1 position on aspartic acid providing enhanced stability and increased cell permeability. The Z-LEHD-FMK inhibitor can be used for both in vivo and in vitro cell based assays to inhibit primarily caspase-9 activity and to study events downsteam of caspase-9 activation.

  • Contents: Lyophilized; Reconstitute in DMSO
  • Format: Purified
  • Molecular weight: 804 Daltons

Avoid multiple freeze-thaws of product.

Flow cytometry

TRUSTED_SUSTAINABILITY

Spécification

For Use With (Application) Flow cytometry
Storage Buffer Lyophilized in Dimethyl Sulfoxide
Content And Storage -20°C
Quantity 1 mg
Molecular Weight (g/mol) 804
Inhibitors Z-LEHD-FMK
Form Purified

For research use only.

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.