missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Includes
Reagents for capture and detection of IL-2, IL-4, IL-6, IL-10, TNF, and IFN-γ; wash buffer; assay diluent; serum enhancement buffer; cytometer setup reagents
- Can be used to quantitatively measure Interleukin-2 (IL-2), Interleukin-4 (IL-4), Interleukin-6 (IL-6), Interleukin-10 (IL-10), Tumor Necrosis Factor (TNF), and Interferon-γ (IFN-γ) protein levels in a single sample
- Detection reagent provided in the kit is a mixture of phycoerythrin (PE)-conjugated antibodies, which provides a fluorescent signal in proportion to the amount of bound analyte
- Provides sufficient reagents for the quantitative analysis of 80 samples
For research use only. Not for use in diagnostic or therapeutic procedures.
Specifications
Specifications
| Detects | IL-2, IL-4, IL-6, IL-10, TNF and IFN-g |
| For Use With (Application) | For analysis of specific cytokines in Tissue Culture supernatants, EDTA Plasma and Serum samples |
| Quantity | 80 Tests |
| Content And Storage | 2°C to 8°C |
| Reactivity | Human |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?