missing translation for 'onlineSavingsMsg'
Learn More

BD CBA Mouse Th1/Th2/Th17 Cytokine Kit

Product Code. 15577135
Change view
Click to view available options
Quantity:
80 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15577135 80 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15577135 Supplier BD Supplier No. 560485
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Reagents for capture and detection of IL-2, IL-4, IL-6, IFN-γ, TNF, IL-17A, and IL-10; wash buffer; assay diluent; cytometer setup reagents

Optimized for analysis of physiologically relevant concentrations (pg/mL levels) of specific cytokine proteins in tissue culture supernatants and serum samples

BD CBA assays provide a method of capturing a soluble analyte or set of analytes with beads of known size and fluorescence, making it possible to detect analytes using flow cytometry.

  • Can be used to measure Interleukin-2 (IL-2), Interleukin-4 (IL-4), Interleukin-6 (IL-6), Interferon-γ (IFN-γ), Tumor Necrosis Factor (TNF), Interleukin-17A (IL-17A), and Interleukin-10 (IL-10) protein levels in a single sample
  • Provides sufficient reagents for 80 tests

TRUSTED_SUSTAINABILITY

Specifications

Detects IL-2, IL-4, IL-6, IFN-g, TNF, IL-17A and IL-10
For Use With (Application) For analysis of physiologically relevant concentrations (pg/mL levels) of specific Cytokine Proteins in Tissue Culture supernatants and Serum samples
Quantity 80 Tests
Content And Storage 2°C to 8°C
compliance-icons
Product Identifier
  • Mouse Th1/Th2/Th17 CBA Kit
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: CMIT/MIT

For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.