missing translation for 'onlineSavingsMsg'
Learn More

BD Cell Detachment Solution

Product Code. 15811109
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15811109 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15811109 Supplier BD Supplier No. 561527
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Replacement for trypsin solution and can be used for the passaging of cell lines

  • Does not contain mammalian or bacterial derived products
  • Comprised of collagenolytic and proteolytic enzymes
  • Performs well when detaching cells for the analysis of many cell surface markers using flow cytometry and for cell sorting

Flow Cytometry, Cell Culture

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Product Type Cell Culture Dissociation Reagent
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.